Learn More
Invitrogen™ KANSL1 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.
Specifications
Specifications
| Antigen | KANSL1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | KANSL1 |
| Gene Accession No. | Q7Z3B3 |
| Gene Alias | 1700081L11Rik; 9430041J06Rik; CENP-36; centromere protein 36; centromere protein 36 antibody; DKFZp686P06109 antibody; DKFZp727C091 antibody; hMSL1v1; Kansl1; KAT8 regulatory NSL complex subunit 1; KAT8 regulatory NSL complex subunit 1 antibody; KDVS; KIAA1267; KIAA1267 antibody; male-specific lethal 1 homolog; MGC102843 antibody; mKIAA1267; MLL1/MLL complex subunit KANSL1; MLL1/MLL complex subunit male-specific lethal 1 homolog antibody; MSL1 homolog 1; MSL1V1; MSL1v1 antibody; non-specific lethal 1 homolog; NSL complex protein NSL1; NSL1; RGD1311429 |
| Gene Symbols | KANSL1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.