Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KANSL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585459
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585459 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585459 Supplier Invitrogen™ Supplier No. PA585459
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (89%). Positive Control: THP-1 whole cell extract, THP-1 nuclear extract. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a nuclear protein that is a subunit of two protein complexes involved with histone acetylation, the MLL1 complex and the NSL1 complex. The corresponding protein in Drosophila interacts with K(lysine) acetyltransferase 8, which is also a subunit of both the MLL1 and NSL1 complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KANSL1
Applications Western Blot
Classification Polyclonal
Concentration 0.82 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene KANSL1
Gene Accession No. Q7Z3B3
Gene Alias 1700081L11Rik; 9430041J06Rik; CENP-36; centromere protein 36; centromere protein 36 antibody; DKFZp686P06109 antibody; DKFZp727C091 antibody; hMSL1v1; Kansl1; KAT8 regulatory NSL complex subunit 1; KAT8 regulatory NSL complex subunit 1 antibody; KDVS; KIAA1267; KIAA1267 antibody; male-specific lethal 1 homolog; MGC102843 antibody; mKIAA1267; MLL1/MLL complex subunit KANSL1; MLL1/MLL complex subunit male-specific lethal 1 homolog antibody; MSL1 homolog 1; MSL1V1; MSL1v1 antibody; non-specific lethal 1 homolog; NSL complex protein NSL1; NSL1; RGD1311429
Gene Symbols KANSL1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human KIAA1267. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284058
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.