Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KAO Monoclonal Antibody (C3), Invitrogen™
GREENER_CHOICE

Catalog No. FEMA551037
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551037 200 μL
FEMA551038 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA551037 Supplier Invitrogen™ Supplier No. MA551037
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This gene encodes a metal-binding membrane glycoprotein that oxidatively deaminates putrescine, histamine, and related compounds. The encoded protein is inhibited by amiloride, a diuretic that acts by closing epithelial sodium ion channels. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KAO
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene AOC1
Gene Accession No. P19801, Q9TRC7
Gene Alias 1600012D06Rik; Abp; abp1; amiloride binding protein 1 (amine oxidase (copper-containing)); amiloride binding protein 1 (amine oxidase, copper-containing); amiloride binding protein/diamine oxidase; amiloride-binding protein 1; amiloride-sensitive amine oxidase; amiloride-sensitive amine oxidase [copper-containing]; amine oxidase copper containing 1; Amine oxidase copper domain-containing protein 1; amine oxidase, copper containing 1; amine oxidase, copper-containing 1; aoc1; DAO; DAO1; Diamine oxidase; histaminase; KAO; kidney amine oxidase; si:ch211-286c5.2; zgc:154101
Gene Symbols AOC1
Host Species Mouse
Immunogen Recombinant Human ABP1 protein, Thr400-Val751 (Accession #P19801).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100517436, 26
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.