Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KAP1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500159AF647
Description
KAP1 Polyclonal antibody specifically detects KAP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
KAP1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
E3 SUMO-protein ligase TRIM28, EC 6.3.2.-, FLJ29029, KAP-1, KAP1KRAB-associated protein 1, KRIP-1, Nuclear corepressor KAP-1, RNF96KRAB-interacting protein 1, TF1B, TIF1-beta, TIF1BRING finger protein 96, transcription intermediary factor 1-beta, transcriptional intermediary factor 1-beta, tripartite motif containing 28, tripartite motif-containing 28, Tripartite motif-containing protein 28 | |
The immunogen recognized by this antibody maps to a region between residue 650 and 700 of human Tripartite Motif-Containing 28 using the numbering given in entry NP_005753.1 (GeneID 10155). | |
0.1 mL | |
Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators, Zinc Finger | |
10155 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction