Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KAP1 Antibody (BL-248-2G6), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP276411
Description
KAP1 Monoclonal specifically detects KAP1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.Specifications
KAP1 | |
Monoclonal | |
Unconjugated | |
Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA with 0.09% Sodium Azide | |
TRIM28 | |
Surrounding serine 824 | |
100 μL | |
10155.0 | |
Store at 2-8°C/ 1 year from date of receipt | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
BL-248-2G6 | |
Western Blot 1:1000, Flow Cytometry, Immunohistochemistry 1:100 - 1:500, Immunoprecipitation 20 ul/mg lysate, Immunocytochemistry 1:100 - 1:500 | |
E3 SUMO-protein ligase TRIM28, EC 6.3.2.-, FLJ29029, KAP-1, KAP1KRAB-associated protein 1, KRIP-1, Nuclear corepressor KAP-1, RNF96KRAB-interacting protein 1, TF1B, TIF1-beta, TIF1BRING finger protein 96, transcription intermediary factor 1-beta, transcriptional intermediary factor 1-beta, tripartite motif containing 28, tripartite motif-containing 28, Tripartite motif-containing protein 28 | |
Rabbit | |
Antigen Affinity-purified | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction