Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KAP1, p Ser824 Antibody, R&D Systems™
SDP

Catalog No. NB1002350 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002350 Supplier R&D Systems Supplier No. NB1002350

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

KAP1 (p Ser824) Polyclonal specifically detects KAP1 (p Ser824) in Human, Mouse, Rat samples. It is validated for Chemotaxis, ChIP Assay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen KAP1 (p Ser824)
Applications Chemotaxis, ChIP Assay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunohistochemistry 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunoprecipitation 2-10 ug/mg lysate, Immunohistochemistry-Paraffin 1:500 - 1:2000, Chromatin Immunoprecipitation (ChIP)
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q13263
Gene Alias E3 SUMO-protein ligase TRIM28, EC 6.3.2.-, FLJ29029, KAP-1, KAP1KRAB-associated protein 1, KRIP-1, Nuclear corepressor KAP-1, RNF96KRAB-interacting protein 1, TF1B, TIF1-beta, TIF1BRING finger protein 96, transcription intermediary factor 1-beta, transcriptional intermediary factor 1-beta, tripartite motif containing 28, tripartite motif-containing 28, Tripartite motif-containing protein 28
Gene Symbols TRIM28
Host Species Rabbit
Immunogen Immunogen was a synthetic phosphorylated peptide, which represented a portion of human KRAB-Associated Protein 1 surrounding Serine 824 according to the numbering given in entry NP_005753.1 (GeneID 10155).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 10155
Test Specificity Mouse reactivity reported in scientific literature (PMID: 24248351) Expected to cross react based on 100% sequence identity: Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque and Northern white-cheeked gibbon.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C. Do not freeze.
Product Type Antibody
Isotype IgG
Product Line Novus
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.