Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KAP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554876
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554876 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554876 Supplier Invitrogen™ Supplier No. PA554876
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSSKPKDPPP FEGMEIDEVA NINDMDEYIE LLYEDIPDKV RGSALILQLA RNPDNLEELL LNETALGALA RVLREDWKQS VELATNIIYI F Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 95%.

The small G protein GDP dissociation stimulator (smg GDS) is a regulator protein having two activities on a group of small G proteins including the Rho and Rap1 family members and Ki-Ras; one is to stimulate their GDP/GTP exchange reactions, and the other is to inhibit their interactions with membranes. The protein encoded by this gene contains 9 'Armadillo' repeats and interacts with the smg GDS protein through these repeats. This protein, which is highly concentrated around the endoplasmic reticulum, is phosphorylated by v-src, and this phosphorylation reduces the affinity of the protein for smg GDS. It is thought that this protein serves as a linker between human chromosome-associated polypeptide (HCAP) and KIF3A/B, a kinesin superfamily protein in the nucleus, and that it plays a role in the interaction of chromosomes with an ATPase motor protein. Several transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KAP3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIFAP3
Gene Accession No. Q92845
Gene Alias dJ190I16.1; FLA3; KAP-1; KAP3; KAP-3; KIF3AP; Kifap3; kinesin associated protein 3; kinesin-associated protein 3; small G protein GDP dissociation stimulator; SMAP; Smg GDS-associated protein; Smg-GDS
Gene Symbols KIFAP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KAP3. Recombinant protein control fragment (Product #RP-94022).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22920
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.