Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KAP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119731
This item is not returnable.
View return policy
Description
Positive Control: K562 cell lysate, rat testis tissue lysate, mouse hippocampus tissue lysates, rat bladder tissue, human liver carcinoma tissue, human thyroid tissue, SHSY5Y. Subcellular Location: cytosol, endoplasmic reticulum, Golgi apparatus, Nucleus.
The small G protein GDP dissociation stimulator (smg GDS) is a regulator protein having two activities on a group of small G proteins including the Rho and Rap1 family members and Ki-Ras; one is to stimulate their GDP/GTP exchange reactions, and the other is to inhibit their interactions with membranes. The protein encoded by this gene contains 9 'Armadillo' repeats and interacts with the smg GDS protein through these repeats. This protein, which is highly concentrated around the endoplasmic reticulum, is phosphorylated by v-src, and this phosphorylation reduces the affinity of the protein for smg GDS. It is thought that this protein serves as a linker between human chromosome-associated polypeptide (HCAP) and KIF3A/B, a kinesin superfamily protein in the nucleus, and that it plays a role in the interaction of chromosomes with an ATPase motor protein. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
| KAP3 | |
| Polyclonal | |
| Unconjugated | |
| KIFAP3 | |
| dJ190I16.1; FLA3; KAP-1; KAP3; KAP-3; KIF3AP; Kifap3; kinesin associated protein 3; kinesin-associated protein 3; small G protein GDP dissociation stimulator; SMAP; Smg GDS-associated protein; Smg-GDS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16579, 22920, 289168 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| P70188, Q92845 | |
| KIFAP3 | |
| Recombinant protein within human KIFAP3 aa 1-200. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction