Learn More
Kappa Light Chain Ab-2 Rabbit Polyclonal Antibody, Epredia™
Recommended for Immunohistochemistry (Formalin/paraffin), Epredia™ Kappa Light Chain Ab-2, Rabbit Polyclonal Antibody is one marker in a comprehensive antibody portfolio that addresses all major disease states, ensuring the ideal tests are always available.
Supplier: Epredia RB333A1
Description
Antibody to the κ light chain of immunoglobulin is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Baboon, Monkey and Rat. Does not react with dog. Others not known.
Immunogen: Kappa light chains purified from human Bence Jones proteins
Molecular Weight: 25kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Kappa Light Chain Ab-2 | |
Polyclonal | |
Unconjugated | |
kappa light chains purified from human Bence Jones proteins | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
Polyclonal | |
Rabbit | |
500 μL | |
Hematopoietic | |
Human, Non-human Primate | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.