Learn More
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Baboon, Monkey and Rat. Does not react with dog. Others not known.
Immunogen: Kappa light chains purified from human Bence Jones proteins
Molecular Weight: 25kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Kappa Light Chain Ab-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Clone | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | kappa light chains purified from human Bence Jones proteins |
| Quantity | 12 mL |
| Regulatory Status | IVD |
| Research Discipline | Hematopoietic |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.