Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kappa Light Chain Antibody (HP6053), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234639G
Description
Kappa Light Chain Monoclonal specifically detects Kappa Light Chain in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Kappa Light Chain | |
Monoclonal | |
DyLight 488 | |
HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
Mouse | |
22.5 kDa | |
0.1 mL | |
Adaptive Immunity, Immunology | |
3514 | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
HP6053 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
IGKC | |
Purified human Ig kappa chain | |
Protein A or G purified | |
RUO | |
Primary | |
This monoclonal antibody is specific to kappa light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of Kappa to Lambda is 70:30. However, with the occurrence of multiple myeloma or other B-cell malignancies this ratio is disturbed. Antibody to the kappa light chain is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is malignant. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction