Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kappa Light Chain Antibody (Kap-56), Allophycocyanin , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311599APC
Description
Kappa Light Chain Monoclonal antibody specifically detects Kappa Light Chain in Human samples. It is validated for Flow CytometrySpecifications
Kappa Light Chain | |
Monoclonal | |
APC | |
HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
Human lymphocytes stimulated | |
0.1 mL | |
Adaptive Immunity, Immunology | |
3514 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry | |
Kap-56 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction