Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kappa Light Chain Antibody (MEM-09), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500331PECY7
Description
Kappa Light Chain Monoclonal specifically detects Kappa Light Chain in Human samples. It is validated for Flow Cytometry.Specifications
Kappa Light Chain | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
IGKC | |
Crude thymus membrane fraction. | |
0.1 mL | |
Primary | |
The antibody MEM-09 reacts with both secreted and B cell-surface human immunoglobulin, specifically reacts with kappa light chains (22.5 kDa). Material immunoprecipitated from human serum with the antibody MEM-09 consists of IgG and traces of IgM. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
MEM-09 | |
Flow Cytometry | |
HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
Mouse | |
Protein A purified | |
Adaptive Immunity, Immunology | |
3514.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction