Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kappa Light Chain Mouse anti-Human, Clone: KLC1278, Abnova™

Catalog No. 89034647 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-647 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-647 Supplier Abnova Corporation Supplier No. MAB14564
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against full length recombinant human Kappa Light Chain .

  • High specificity
  • High reactivity

Specifications

Antigen Kappa Light Chain
Applications Flow Cytometry, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone KLC1278
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human Kappa Light Chain .
Dilution Flow Cytometry (0.5-1 ug/106 cells) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Gene Accession No. P01834
Gene Alias HCAK1/Km/MGC111575/MGC62011/MGC72072/MGC88770/MGC88771/MGC88809
Gene Symbols IGKC
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human Kappa Light Chain .
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3514
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.