Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kappa Light Chain Antibody (KLC1278) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$610.00
Specifications
Antigen | Kappa Light Chain |
---|---|
Clone | KLC1278 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254512100
![]() |
Novus Biologicals
NBP254512100UG |
100 μg |
Each for $610.00
|
|
|||||
NB254512200
![]() |
Novus Biologicals
NBP254512200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Kappa Light Chain Monoclonal specifically detects Kappa Light Chain in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
Kappa Light Chain | |
1 mg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Immunology | |
PBS with No Preservative | |
3514 | |
Recombinant human full-length Ig kappa chain | |
Primary | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
KLC1278 | |
Flow Cytometry 0.5 - 1 ug/million cells, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
IGKC | |
IgG1 κ | |
Protein A or G purified | |
22.5 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title