Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kappa light chain Rabbit anti-Mouse, Clone: RM103, Abnova™
Rabbit monoclonal antibody raised against native mouse immunoglobulin kappa light chain.
Supplier: Abnova Corporation MAB12764
Specifications
Kappa light chain | |
Monoclonal | |
Unconjugated | |
ELISA (0.005 - 0.2 ug/mL) Flow Cytometry Immunocytochemistry (0.5 - 2 ug/mL) Immunohistochemistry (0.5 - 2 ug/mL) Immunoprecipitation Western Blot (0.1 - 0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
IGKC/MGC118128 | |
Rabbit | |
Protein A affinity purification | |
RUO | |
16071 | |
Mouse | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
RM103 | |
Rabbit monoclonal antibody raised against native mouse immunoglobulin kappa light chain. | |
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
Igk-C | |
Native purified IgG from mouse serum. | |
100 μg | |
Primary | |
This antibody reacts to the kappa light chain of mouse immunoglobulins. No cross reactivity with the lambda light chain, human IgG, rat IgG, or goat IgG. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction