Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KappaB ras1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103496
Description
Antibody detects endogenous levels of total NKIRAS1.
KappaB ras-1 (kappa-B-ras-1) and kappaB-ras-2 are two small proteins that similar to Ras-like small GTPases that associate with Ikappa-B (Ikappa-B), an inhibitor of the transcription factor NF-kappa-B. Ikappa-B exists in two homologous forms, Ikappa-B-alpha and Ikappa-B-beta, although Ikappa-B-beta contains a unique 47-amino acid region within its ankyrin domain. While inactive Ikappa-B-alpha-NF-kappa-B complexes can shuttle in and out of the nucleus, Ikappa-B-beta-NF-kappa-B complexes are retained exclusively in the cytoplasm. It is suggested that kappaB-ras proteins preferentially bind to the Ikappa-B-beta form through this unique insert within the ankyrin region, thus modulating the cellular location of Ikappa-B-beta and regulating the rate of degradation of Ikappa-B-beta.
Specifications
| KappaB ras1 | |
| Polyclonal | |
| Unconjugated | |
| NKIRAS1 | |
| 2400004O09Rik; C15orf1; I1PP2A; I-kappa-B-interacting Ras-like protein 1; kappa B-ras 1; kappa B-Ras protein 1; kappaB-Ras1; KBRAS1; LANP; MAPM; MGC119787; MGC150373; NF-kappa-B inhibitor-interacting Ras-like protein 1; NFKB inhibitor interacting Ras like 1; NFKB inhibitor interacting Ras-like 1; NFKB inhibitor interacting Ras-like 1 S homeolog; NFKB inhibitor interacting Ras-like protein 1; Nkiras1; nkiras1.S; PHAP1; PHAPI; PP32; XELAEV_18033117mg | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 28512, 69721 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8CEC5, Q9NYS0 | |
| NKIRAS1 | |
| A synthesized peptide derived from human NKIRAS1(Accession Q9NYS0), corresponding to amino acid residues D153-N192. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction