Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KappaB ras2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA520342

Catalog No. PIPA520342


Only null left
Add to Cart

Description

Description

A suggested positive control is RAW264.7 cell lysate. PA5-20342 can be used with blocking peptide PEP-0462.

Atypical Ras-like protein that acts as a potent regulator of NF-kappa-B activity by preventing the degradation of NF-kappa-B inhibitor beta (NFKBIB) by most signals, explaining why NFKBIB is more resistant to degradation. May act by blocking phosphorylation of NFKBIB and nuclear localization of p65/RELA NF-kappa-B subunit. It is unclear whether it acts as a GTPase. Both GTP- and GDP-bound forms block phosphorylation of NFKBIB.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KappaB ras2
Polyclonal
Unconjugated
NKIRAS2
2410003M04Rik; 4930527H08Rik; D630018G21Rik; I-kappa-B-interacting Ras-like protein 2; Kappa B-Ras protein 2; kappaB-Ras2; KBRAS2; NF-kappa-B inhibitor-interacting Ras-like protein 2; NFKB inhibitor interacting Ras like 2; NFKB inhibitor interacting Ras-like 2; NFKB inhibitor interacting Ras-like protein 2; NKIRAS2
Rabbit
Antigen affinity chromatography
RUO
28511, 287707, 71966
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9CR56, Q9NYR9
NKIRAS2
A 14 amino acid peptide from near the carboxy terminus of human KappaB ras2.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.