Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kappler™ Frontline 300 Chemical and Fire Protection Coverall with Hood Opening
Shop All Kappler, Inc. Products

Click to view available options
Size:
Small/Medium, Medium
Large/X-Large, X-Large
2X-Large/3X-Large, 3X-Large
4X-Large
5X-Large
Specifications
Specifications
Size | Small/Medium, Medium |
Material | Frontline™ |
Color | Silver |
Ankle Style | Attached Boots, Boot Overflaps |
Cuff Style | Attached Gloves |
Closure Type | Double Storm Flap with Hook and Loop Seal |
Seam Type | Bound, Piped, Sealed |
Resistance | Flame, Chemicals |
Back Style | Expanded |
Certifications/Compliance | ASTM F1001 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction