Kappler™ ProVent™ 7000 Isolation Gowns

Resistant to abrasions, punctures, and tears

Manufacturer: Kappler, Inc.Supplier Diversity Partner LS112WH3X

 View more versions of this product

Catalog No. 18-025-653

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) For blood penetration resistance
    Size 3X-Large
    Sterile No
    Weight (English) 17 lb.
    Weight (Metric) 7.7kg
    Cuff Style Elastic
    Certifications/Compliance ASTM 1670
    Color White
    Material Composite Fabric

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit