Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Karl Fischer Coulometric CG, catholyte solution, Thermo Scientific™

Catalog No. p-7052264
Click to view available options
Quantity:
6 x 110 mL

Catalog No. AA47117RDL

Supplier: Thermo Scientific Chemicals 47117RDL

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Karl Fischer Coulometric CG
Name Note Catholyte solution
Boiling Point 202°C
Color Colorless to Yellow
Density 1.1 g/mL
Physical Form Liquid
Quantity 6 x 110 mL
Flash Point 92°C (198°F)
UN Number UN3267
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.