Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KARS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559620
Description
Immunogen sequence: GLTERFELFV MKKEICNAYT ELNDPMRQRQ LFEEQAKAKA AGDDEAMFID ENFCTALEYG LPPTAGWGMG IDRVAMFLTD SNNIKEVLLF PAMKPEDKKE NVATTDTLES T Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. Lysyl-tRNA synthetase is a homodimer localized to the cytoplasm which belongs to the class II family of tRNA synthetases. It has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
KARS | |
Polyclonal | |
Unconjugated | |
KARS1 | |
AA589550; AL024334; AL033315; AL033367; CMTRIB; D8Ertd698e; D8Wsu108e; DFNB89; EC 6.1.1.6; KARS; Kars1; KARS2; KIAA0070; KRS; lysine tRNA ligase; lysine--tRNA ligase; LysRS; lysyl-tRNA synthetase; mKIAA0070 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
292028, 3735, 85305 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15046, Q5XIM7, Q99MN1 | |
KARS1 | |
Recombinant protein corresponding to Human KARS. Recombinant protein control fragment (Product #RP-98537). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction