Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KARS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589426
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589426 Supplier Invitrogen™ Supplier No. PA589426
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IAPELYHKML VVGGIDRVYE IGRQFRNEGI DLTHNPEFTT CEFYMAYADY HDLMEITEKM VSGMVKHITG SYKVTYHPDG PEGQAYDVDF TPPFRRINMV EELEKALGMK LPETNLFETE ETRKILDDIC VAKAVECPPP RTTARLLDKL VGEFLEVTCI NPTFICDHPQ IMSPLAKWHR SKEGLTERFE LFVMKKEICN AYTELNDPMR QRQLFEEQAK AKAAGDDEAM FIDENFCTAL EYGLPPTAGW GMGIDRVAMF LTDSNNIKEV LLFPAMKPED KKENVATTDT LESTTVGTSV; Positive Samples: HepG2, Jurkat, HeLa, 293T, Mouse heart, Mouse kidney, Rat liver; Cellular Location: Cell membrane, Cytoplasm, Mitochondrion, Nucleus, Peripheral membrane protein.

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. Lysyl-tRNA synthetase is a homodimer localized to the cytoplasm which belongs to the class II family of tRNA synthetases. It has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KARS
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KARS1
Gene Accession No. Q15046, Q5XIM7, Q99MN1
Gene Alias AA589550; AL024334; AL033315; AL033367; CMTRIB; D8Ertd698e; D8Wsu108e; DFNB89; EC 6.1.1.6; KARS; Kars1; KARS2; KIAA0070; KRS; lysine tRNA ligase; lysine--tRNA ligase; LysRS; lysyl-tRNA synthetase; mKIAA0070
Gene Symbols KARS1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 298-597 of human LysRS/LysRS/KARS (NP_0055391).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 292028, 3735, 85305
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.