Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Kartogenin

Catalog No. 451310
Change view
Click to view available options
Quantity:
10 mg
50 mg
Catalog No. Quantity
45-131-0 10 mg
45-135-0 50 mg
2 options
This item is not returnable. View return policy

Catalog No. 45-131-0

Supplier: Tocris Bioscience™ 4513/10

This item is not returnable. View return policy

Potently induces chondrogenesis in MSCs

Kartogenin potently induces differentiation of human mesenchymal stem cells into chondrocytes (EC50 = 100 nM). Reduces disease severity in a mouse model of osteoarthritis; displays protective effects against osteoarthritic stimuli in mature chondrocytes in vitro.

Chemical Identifiers

CAS 4727-31-5
Molecular Formula C20H15NO3

Specifications

Quantity 10 mg
Formula Weight 317.34
Purity >98%
Molecular Weight (g/mol) 317.344
Chemical Name or Material Kartogenin
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.