Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KAT IIRabbit anti-Mouse, Polyclonal, Rockland™
SDP

Catalog No. 501992204
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2204 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-2204 Supplier Rockland Immunochemicals Supplier No. 600401J09
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. A BLAST analysis was used to suggest cross-reactivity with human, rat, and mouse based on 100% sequence homology. Cross-reactivity with KAT II from other sources has not been determined.

This gene encodes a protein that is highly similar to mouse and rat kynurenine aminotransferase II. The rat protein is a homodimer with two transaminase activities. One activity is the transamination of alpha-aminoadipic acid, a final step in the saccaropine pathway which is the major pathway for L-lysine catabolism. The other activity involves the transamination of kynurenine to produce kynurenine acid, the precursor of kynurenic acid which has neuroprotective properties. Two alternative transcripts encoding the same isoform have been identified, however, additional alternative transcripts and isoforms may exist.

Specifications

Antigen KAT II
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.05 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl, 30% glycerol and no preservative; pH 7.2
Gene Aadat
Gene Accession No. Q9WVM8
Gene Alias 2-aminoadipate aminotransferase; 2-aminoadipate transaminase; AADAT; Aadt; AI875679; Alpha-aminoadipate aminotransferase; aminoadipate aminotransferase; KAT/AadAT; Kat2; KATII; KYAT2; kynurenine aminotransferase 2; Kynurenine aminotransferase II; Kynurenine/alpha-aminoadipate aminotransferase, mitochondrial; kynurenine--oxoglutarate; Kynurenine--oxoglutarate aminotransferase II; Kynurenine--oxoglutarate transaminase 2; Kynurenine--oxoglutarate transaminase II; L kynurenine/alpha aminoadipate aminotransferase; mKat-2
Gene Symbols Aadat
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23923
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.