Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KAT1 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298804MFV500
Description
KAT1 Polyclonal antibody specifically detects KAT1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
KAT1 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
EC 2.3.1.48, histone acetyltransferase 1, histone acetyltransferase type B catalytic subunit, KAT1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human KAT1. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
8520 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction