Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KAT2A/GNC5 Mouse anti-Human, Mouse, Rat, Clone: 1F6G7, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | KAT2A/GNC5 |
---|---|
Clone | 1F6G7 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6993
![]() ![]() |
Proteintech Group Inc
665751IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
SAGA (Spt-Ada-Gen5 histone acetyltransferase complex) is a histoneacetylase complex which has Gcn5p as catalyst subunit and functions overlapping with the fundamental transcription factor TFIID which has Taf1p as catalyst subunit. However, SAGA and TFIID have different allotment and each accomplishes the important role in the transcription for Housekeeping gene group and Stress Responding gene group. Also, SAGA is a gigantic protein complex which is composed of Ada protein group (5 kinds), TBP related protein group (4 kinds), TAF protein group (5 kinds that also pertain to TFIID), and other protein groups (>6 kinds). As its molecular function, there is the chemical modification of histone, or the recruitment by direct interaction of transcription regulating factor on DNA, or the control of transcription starting reaction by TBP. Gcn5p that show histoneacetylase activity is one kind of the above-mentioned Ada protein group, and in the case of budding yeast it is composed of 439 amino acid residues.Specifications
KAT2A/GNC5 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
GCN5, GCN5L2, hGCN5, Histone acetyltransferase GCN5, HsGCN5, K(lysine) acetyltransferase 2A, KAT2A, Lysine acetyltransferase 2A, PCAF b, STAF97 | |
Kat2a | |
IgG2a | |
Protein A | |
Antibody |
1F6G7 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
Q92830, Q9JHD2 | |
14534, 2648, 303539 | |
KAT2A/GNC5 Fusion Protein Ag6940 | |
Primary | |
-20°C | |
KAT2A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title