Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KAT3B/p300 Mouse anti-Human, Mouse, Rat, Mustelid, Primate, APC, Clone: RW105, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100616APC0.1ML
This item is not returnable.
View return policy
Description
KAT3B/p300 Monoclonal antibody specifically detects KAT3B/p300 in Human, Mouse, Rat, Mustelid, Primate samples. It is validated for Flow (Intracellular)Specifications
KAT3B/p300 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Mustelid, Primate | |
IgG1 κ |
Flow Cytometry | |
RW105 | |
Flow Cytometry | |
E1A binding protein p300, E1A-associated protein p300, E1A-binding protein, 300kD, EC 2.3.1, EC 2.3.1.48, histone acetyltransferase p300, KAT3B, P300, p300 HAT, RSTS2 | |
Fusion protein containing residues 1572-2371 of human KAT3B/p300. [UniProt# Q0947] | |
0.1 mL | |
Adaptive Immunity, Alzheimers Research, Amino Acids Drugs and other small molecules, Angiogenesis, Apoptosis, Asthma, Autophagy, Base Excision Repair, Beta Galactosidase Epitope Tags, Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors, Vision, Wnt Signaling Pathway | |
2033 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction