Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KAT4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984985
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4985 Supplier Bioss Supplier No. BS2812R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

3-Phosphoglycerate dehydrogenase (PHGDH; EC 1.1.1.95) catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.

Specifications

Antigen KAT4
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TAF1
Gene Accession No. P21675
Gene Alias AU015687; B430306D02Rik; BA2R; Ccg1; Ccg-1; CCGS; Cell cycle gene 1 protein; cell cycle, G1 phase defect; complementation of cell cycle block, G1-to-S; DYT3; DYT3/TAF1; KAT4; MRXS33; NSCL2; N-TAF1; OF; p250; RGD1562050; TAF(II)250; TAF1; TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 250kDa; TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, neuron specific isoform; TAF2A; TAFII250; TAFII-250; TATA box binding protein associated factor 1; TATA-box binding protein associated factor 1; TBP-associated factor 250 kDa; transcription factor TFIID p250 polypeptide; Transcription initiation factor TFIID 250 kDa subunit; Transcription initiation factor TFIID subunit 1; XDP
Gene Symbols TAF1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human TBP Associated Factor 1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6872
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.