Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KAT5 Mouse anti-Human, Clone:4B3B4, Abnova™

Catalog No. 89333167 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-167 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-167 Supplier Abnova Corporation Supplier No. MAB16675
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human KAT5.

The protein encoded by this gene belongs to the MYST family of histone acetyl transferases (HATs) and was originally isolated as an HIV-1 TAT-interactive protein. HATs play important roles in regulating chromatin remodeling, transcription and other nuclear processes by acetylating histone and nonhistone proteins. This protein is a histone acetylase that has a role in DNA repair and apoptosis and is thought to play an important role in signal transduction. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq]

Specifications

Antigen KAT5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4B3B4
Conjugate Unconjugated
Description K(lysine) acetyltransferase 5
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry
Flow Cytometry
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene KAT5
Gene Alias ESA1, HTATIP, HTATIP1, PLIP, TIP, TIP60, cPLA2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 18-208 of human KAT5 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 10524
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.