Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Katanin p60 Rabbit anti-Human, Clone: S05-3B9, Novus Biologicals™

Rabbit Monoclonal Antibody
$254.41 - $556.36
Specifications
| Antigen | Katanin p60 |
|---|---|
| Clone | S05-3B9 |
| Dilution | Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Monoclonal |
Description
Katanin p60 Monoclonal antibody specifically detects Katanin p60 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
| Katanin p60 | |
| Western Blot, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.6.4.3, katanin p60 (ATPase containing) subunit A 1, katanin p60 (ATPase-containing) subunit A 1, katanin p60 ATPase-containing subunit A1, Katanin p60 subunit A1, p60 katanin | |
| A synthetic peptide of human Katanin p60 (Uniprot#: O75449) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S05-3B9 | |
| Western Blot, ELISA, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 11104 | |
| IgG | |
| Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title