Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Katanin p80 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559462
Description
Immunogen sequence: SPAMDVQFPV PNLEVLPRPP VVASTPAPKA EPAIIPATRN EPIGLKASDF LPAVKIPQQA ELVDEDAMSQ IRKGHDTMCV VLTSRHKNLD TVRAVWTMGD IKTSVDSAVA INDLSVV Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.
Microtubules, polymers of alpha and beta tubulin subunits, form the mitotic spindle of a dividing cell and help to organize membranous organelles during interphase. Katanin is a heterodimer that consists of a 60 kDa ATPase (p60 subunit A 1) and an 80 kDa accessory protein (p80 subunit B 1). The p60 subunit acts to sever and disassemble microtubules, while the p80 subunit targets the enzyme to the centrosome. Katanin is a member of the AAA family of ATPases.
Specifications
Katanin p80 | |
Polyclonal | |
Unconjugated | |
Katnb1 | |
2410003J24Rik; KAT; katanin (80 kDa); katanin p80 (WD repeat containing) subunit B 1; katanin p80 (WD40-containing) subunit B 1; katanin p80 subunit B1; Katanin p80 WD40 repeat-containing subunit B1; katanin p80 WD40-containing subunit B1; katanin regulatory subunit B1; KATNB1; LIS6; p80 katanin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10300 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BVA0 | |
Katnb1 | |
Recombinant protein corresponding to Human KATNB1. Recombinant protein control fragment (Product #RP-98570). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction