Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KATNA1 Polyclonal Antibody

Catalog No. PIPA5121810
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121810 20 μL
PIPA5119239 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121810 Supplier Invitrogen™ Supplier No. PA5121810
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KATNA1 Polyclonal Antibody for Western Blot, IHC (P)

Microtubules, polymers of alpha and beta tubulin subunits, form the mitotic spindle of a dividing cell and help to organize membranous organelles during interphase. Katanin is a heterodimer that consists of a 60 kDa ATPase (p60 subunit A 1) and an 80 kDa accessory protein (p80 subunit B 1). The p60 subunit acts to sever and disassemble microtubules, while the p80 subunit targets the enzyme to the centrosome. This gene encodes the p80 subunit. This protein is a member of the AAA family of ATPases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KATNA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene KATNA1
Gene Accession No. O75449, Q9WV86
Gene Alias HAMAP-Rule:MF_03023}; hypothetical protein LOC553631; katanin catalytic subunit A1; katanin p60 (ATPase containing) subunit A 1; katanin p60 (ATPase-containing) subunit A 1; katanin p60 (ATPase-containing) subunit A1; katanin p60 ATPase-containing subunit A1; katanin p60 ATPase-containing subunit A1 {ECO:0000255; Katanin p60 ATPase-containing subunit A1-like protein; katanin p60 subunit A 1; katanin p60 subunit A1; katanin p60 subunit A1 {ECO:0000255; KATNA1; lipotransin; microtubule severing protein; p60 katanin; p60 katanin {ECO:0000255; si:dkey-15j16.1; zgc:110580
Gene Symbols KATNA1
Host Species Rabbit
Immunogen Recombinant protein Katanin P60 Subunit A 1. The antigen corresponds to amino acid range 260-491 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11104, 23924
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.