Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KATNA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569830
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569830 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569830 Supplier Invitrogen™ Supplier No. PA569830
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.

Microtubules, polymers of alpha and beta tubulin subunits, form the mitotic spindle of a dividing cell and help to organize membranous organelles during interphase. Katanin is a heterodimer that consists of a 60 kDa ATPase (p60 subunit A 1) and an 80 kDa accessory protein (p80 subunit B 1). The p60 subunit acts to sever and disassemble microtubules, while the p80 subunit targets the enzyme to the centrosome. This gene encodes the p80 subunit. This protein is a member of the AAA family of ATPases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KATNA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KATNA1
Gene Accession No. Q9WV86
Gene Alias HAMAP-Rule:MF_03023}; hypothetical protein LOC553631; katanin catalytic subunit A1; katanin p60 (ATPase containing) subunit A 1; katanin p60 (ATPase-containing) subunit A 1; katanin p60 (ATPase-containing) subunit A1; katanin p60 ATPase-containing subunit A1; katanin p60 ATPase-containing subunit A1 {ECO:0000255; Katanin p60 ATPase-containing subunit A1-like protein; katanin p60 subunit A 1; katanin p60 subunit A1; katanin p60 subunit A1 {ECO:0000255; KATNA1; lipotransin; microtubule severing protein; p60 katanin; p60 katanin {ECO:0000255; si:dkey-15j16.1; zgc:110580
Gene Symbols KATNA1
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse KATNA1 (aa 347-396).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23924
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.