Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KATNAL1 Antibody (OTI1A7), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272254PE
Description
KATNAL1 Monoclonal specifically detects KATNAL1 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Flow Cytometry.Specifications
KATNAL1 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
KATNAL1 | |
Full length recombinant protein of human KATNAL1 (NP_115492) produced in HEK293T cells. | |
0.1 mL | |
84056.0 | |
Store at 4°C in the dark. |
Flow Cytometry | |
OTI1A7 | |
Flow Cytometry | |
EC 3.6.4.3, katanin p60 ATPase-containing subunit A-like 1, katanin p60 subunit A-like 1MGC2599, p60 katanin-like 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Primate, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction