Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KATNAL1 Monoclonal Antibody (OTI1A7)
GREENER_CHOICE

Catalog No. PIMA525399
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525399 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525399 Supplier Invitrogen™ Supplier No. MA525399
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KATNAL1 Monoclonal Antibody for Western Blot, IHC (P), Flow

KATNAL1 (Katanin Catalytic Subunit A1 Like 1) is a Protein Coding gene. Diseases associated with KATNAL1 include 13Q12.3 Microdeletion Syndrome. Gene Ontology (GO) annotations related to this gene include microtubule binding and microtubule-severing ATPase activity. An important paralog of this gene is KATNA1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KATNAL1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1A7
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KATNAL1
Gene Accession No. Q5XIK7, Q9BW62
Gene Alias HAMAP-Rule:MF_03024}; katanin catalytic subunit A like 1; katanin catalytic subunit A1 like 1; katanin p60 ATPase-containing subunit A-like 1; katanin p60 ATPase-containing subunit A-like 1 {ECO:0000255; katanin p60 subunit A like 1; katanin p60 subunit A-like 1; katanin p60 subunit A-like 1 {ECO:0000255; Katnal1; p60 katanin-like 1; p60 katanin-like 1 {ECO:0000255
Gene Symbols KATNAL1
Host Species Mouse
Immunogen Full length human recombinant protein of KATNAL1 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288449, 84056
Target Species Human, Rat, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.