Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KATNAL1 Monoclonal Antibody (OTI1A7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502344

Description
KATNAL1 (Katanin Catalytic Subunit A1 Like 1) is a Protein Coding gene. Diseases associated with KATNAL1 include 13Q12.3 Microdeletion Syndrome. Gene Ontology (GO) annotations related to this gene include microtubule binding and microtubule-severing ATPase activity. An important paralog of this gene is KATNA1.Specifications
KATNAL1 | |
Monoclonal | |
0.4 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q5XIK7, Q9BW62 | |
Katnal1 | |
Full length human recombit protein of human KATNAL1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1A7 | |
Unconjugated | |
KATNAL1 | |
katanin catalytic subunit A1 like 1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
288449, 84056 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction