Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KATNAL1 Monoclonal Antibody (OTI1A7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502344S


Description
KATNAL1 Monoclonal antibody specifically detects KATNAL1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
KATNAL1 | |
Monoclonal | |
0.4 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P53370 | |
Mouse | |
Affinity chromatography | |
RUO | |
11162 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1A7 | |
Unconjugated | |
NUDT6 | |
HAMAP-Rule:MF_03024}; katanin catalytic subunit A like 1; katanin catalytic subunit A1 like 1; katanin p60 ATPase-containing subunit A-like 1; katanin p60 ATPase-containing subunit A-like 1 {ECO:0000255; katanin p60 subunit A like 1; katanin p60 subunit A-like 1; katanin p60 subunit A-like 1 {ECO:0000255; Katnal1; p60 katanin-like 1; p60 katanin-like 1 {ECO:0000255 | |
Full length human recombinant protein of human NUDT6 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction