Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KATNAL1 Monoclonal Antibody (OTI1F5), TrueMAB™, OriGene
SDP

Catalog No. 502541326
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1326 30 μL
50-167-3783 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1326 Supplier OriGene Supplier No. TA502769S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KATNAL1 Monoclonal antibody specifically detects KATNAL1 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen KATNAL1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1F5
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NFKBIZ
Gene Accession No. Q9BYH8
Gene Alias HAMAP-Rule:MF_03024}; katanin catalytic subunit A like 1; katanin catalytic subunit A1 like 1; katanin p60 ATPase-containing subunit A-like 1; katanin p60 ATPase-containing subunit A-like 1 {ECO:0000255; katanin p60 subunit A like 1; katanin p60 subunit A-like 1; katanin p60 subunit A-like 1 {ECO:0000255; Katnal1; p60 katanin-like 1; p60 katanin-like 1 {ECO:0000255
Host Species Mouse
Immunogen Full length human recombinant protein of human NFKBIZ produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64332
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.