Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KATNAL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531641
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531641 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531641 Supplier Invitrogen™ Supplier No. PA531641
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji, mouse testis. Predicted reactivity: Mouse (95%), Rat (96%), Zebrafish (87%), Rabbit (97%), Rhesus Monkey (98%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

KATNAL1 (Katanin Catalytic Subunit A1 Like 1) is a Protein Coding gene. Diseases associated with KATNAL1 include 13Q12.3 Microdeletion Syndrome. Gene Ontology (GO) annotations related to this gene include microtubule binding and microtubule-severing ATPase activity. An important paralog of this gene is KATNA1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KATNAL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene KATNAL1
Gene Accession No. Q8K0T4, Q9BW62
Gene Alias HAMAP-Rule:MF_03024}; katanin catalytic subunit A like 1; katanin catalytic subunit A1 like 1; katanin p60 ATPase-containing subunit A-like 1; katanin p60 ATPase-containing subunit A-like 1 {ECO:0000255; katanin p60 subunit A like 1; katanin p60 subunit A-like 1; katanin p60 subunit A-like 1 {ECO:0000255; Katnal1; p60 katanin-like 1; p60 katanin-like 1 {ECO:0000255
Gene Symbols KATNAL1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human KATNAL1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231912, 84056
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.