Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KATNAL2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | KATNAL2 |
---|---|
Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
KATNAL2 Polyclonal specifically detects KATNAL2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KATNAL2 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q8IYT4 | |
83473 | |
This antibody was developed against a recombinant protein corresponding to amino acids: NNLPQRSRGKTRRMMNDSCQNLPKINQQRPRSKTTAGKTGDTKSLNKEHPNQEVVDNTRLENANFGLHISRIRK | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
DKFZp667C165, EC 3.6.4.3, katanin p60 ATPase-containing subunit A-like 2, katanin p60 subunit A-like 2DKFZP667C165, MGC33211, p60 katanin-like 2 | |
KATNAL2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title