Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KATNAL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113096
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113096 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113096 Supplier Invitrogen™ Supplier No. PA5113096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KATNAL2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen KATNAL2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene KATNAL2
Gene Accession No. Q8IYT4
Gene Alias katanin catalytic subunit A like 2; katanin catalytic subunit A1 like 2; katanin p60 ATPase-containing subunit A-like 2; katanin p60 subunit A like 2; Katanin p60 subunit A-like 2; KATNAL2; p60 katanin-like 2
Gene Symbols KATNAL2
Host Species Rabbit
Immunogen Recombinant Human Katanin p60 ATPase-containing subunit A-like 2 protein (18-307AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83473
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.