Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KATNB1 Monoclonal Antibody (OTI5A1), TrueMAB™, OriGene
SDP

Catalog No. 502540740
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0740 30 μL
50-167-4548 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0740 Supplier OriGene Supplier No. TA503724S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KATNB1 Monoclonal antibody specifically detects KATNB1 in Canine, Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen KATNB1
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI5A1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SEC14L2
Gene Accession No. O76054, Q99J08
Gene Alias 2410003J24Rik; KAT; katanin (80 kDa); katanin p80 (WD repeat containing) subunit B 1; katanin p80 (WD40-containing) subunit B 1; katanin p80 subunit B1; Katanin p80 WD40 repeat-containing subunit B1; katanin p80 WD40-containing subunit B1; katanin regulatory subunit B1; KATNB1; LIS6; p80 katanin
Host Species Mouse
Immunogen Full length human recombinant protein of human SEC14L2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23541, 477539, 67815
Target Species Canine, Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.