Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KATNB1 Monoclonal Antibody (OTI5H6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503750S


Description
KATNB1 Monoclonal antibody specifically detects KATNB1 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
KATNB1 | |
Monoclonal | |
0.67 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q16576 | |
Mouse | |
Affinity chromatography | |
RUO | |
5931 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI5H6 | |
Unconjugated | |
RBBP7 | |
2410003J24Rik; KAT; katanin (80 kDa); katanin p80 (WD repeat containing) subunit B 1; katanin p80 (WD40-containing) subunit B 1; katanin p80 subunit B1; Katanin p80 WD40 repeat-containing subunit B1; katanin p80 WD40-containing subunit B1; katanin regulatory subunit B1; KATNB1; LIS6; p80 katanin | |
Full length human recombinant protein of human RBBP7 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction