Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KAZALD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119241
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119241 200 μL
PIPA5121808 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119241 Supplier Invitrogen™ Supplier No. PA5119241
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KAZALD1 Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes a secreted member of the insulin growth factor-binding protein (IGFBP) superfamily. It contains an N-terminal insulin growth factor-binding domain, a central Kazal-type serine protease inhibitor and follistatin-like domain, and a C-terminal immunoglobulin-like domain. Studies of the mouse ortholog suggest that this gene product may have a function in bone development and bone regeneration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KAZALD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene KAZALD1
Gene Accession No. Q96I82
Gene Alias bone- and odontoblast-expressed gene 1; BONO1; FKSG28; FKSG40; FLJ24094; IGFBP-related protein 10; IGFBP-rP10; Kazal type serine peptidase inhibitor domain 1; KAZALD1; kazal-type serine protease inhibitor domain-containing protein 1; novel kazal-type serine protease inhibitor domain and immunoglobulin domain containing protein; UNQ2945/PRO21184
Gene Symbols KAZALD1
Host Species Rabbit
Immunogen Recombinant protein Kazal Type Serine Protease Inhibitor Domain Containing Protein 1. The antigen corresponds to amino acid range 31-304 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100624902, 617445, 81621
Target Species Human, Bovine, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.