Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kazrin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989280
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9280 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9280 Supplier Bioss Supplier No. BS9707R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Component of the cornified envelope of keratinocytes. May be involved in the interplay between adherens junctions and desmosomes. The function in the nucleus is not known.

Specifications

Antigen Kazrin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KAZN
Gene Accession No. Q674X7
Gene Alias 2310007B04Rik; 9030409G11Rik; AI256607; AI666764; AW061099; HRIHFB2003; Kaz; KAZN; Kazrin; kazrin, periplakin interacting protein; KIAA1026
Gene Symbols KAZN
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Kazrin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23254
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.