Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ kb NB 142-70
GSA_VAGREENER_CHOICE

Catalog No. 396210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-6210 10 mg
39-625-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-6210 Supplier Tocris Bioscience™ Supplier No. 3962/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective PKD inhibitor; analog of CID 755673 (Cat. No. 3327)

Selective protein kinase D (PKD) inhibitor (IC50 values are 28.3, 58.7 and 53.2 nM for PKD1, 2 and 3 respectively). More potent analog of CID 755673 (Cat. No. 3327) with 7-fold greater inhibition. Inhibits prostrate cancer cell migration and invasion and reduces wound healing in vitro; displays prominent cytotoxic and anti-proliferative effects.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 1233533-04-4
Molecular Formula C11H9NO2S2

Specifications

Quantity 10 mg
Formula Weight 251.32
Purity >98%
Molecular Weight (g/mol) 251.318
Chemical Name or Material kb NB 142-70
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.