Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ KB SRC 4
GREENER_CHOICESDP

Catalog No. 466010 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-601-0 10 mg
46-605-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 46-601-0 Supplier Tocris Bioscience™ Supplier No. 4660/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective c-Src inhibitor

Potent and selective c-Src inhibitor (Ki = 44 nM); does not inhibit c-Abl at concentrations up to 125 μM. Selective between Src family members (Kd values are 86, 160, 240, 720, 3200, 4400, and >40,000 nM for c-Src, Lck, Fgr, Yes, Lyn, Hck and Fyn respectively). Significantly inhibits cell growth in 4T1 mammary carcinoma tumor cells.

Chemical Identifiers

CAS 1380088-03-8
Molecular Formula C32H23ClN8

Specifications

Quantity 10 mg
Formula Weight 555.03
Purity >98%
Molecular Weight (g/mol) 555.042
Chemical Name or Material KB SRC 4
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.