Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KBA.62 (Melanoma Associated Antigen) Monoclonal Antibody (G062.1), QED Bioscience™
SDP

Catalog No. 502278004
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-8004 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-8004 Supplier Leinco Technologies Inc Supplier No. 34062100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

KBA.62 (Melanoma Associated Antigen) Monoclonal antibody specifically detects KBA.62 (Melanoma Associated Antigen) in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen KBA.62 (Melanoma Associated Antigen)
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone G062.1
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation tris with 1% BSA and 0.1% sodium azide; pH 7.3-7.7
Host Species Mouse
Immunogen Recombinant human KBA.62.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.