Learn More
BINDER™ Reinforced Inner Chamber
SDP
Catalog No. 502875633 Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
KB 1060
KB 130
KB 260
KB 470
KB 65
KB 720
KB PRO 1060
KB PRO 130
KB PRO 260
KB PRO 470
KB PRO 720
KBF 1060
KBF 130
KBF 260
KBF 470
KBF 720
KBF PRO 1060
KBF PRO 130
KBF PRO 260
KBF PRO 470
KBF PRO 720
Reinforced inner chamber for climate chambers and cooling incubation chambers.
  • Reinforced inner chamber for bulky and heavy samples.
  • Weight capacity ranged from 100 to 200 kg, depending on model.
Specifications
For Use With (Equipment) KB PRO 720
Material Stainless Steel
Type Chamber Rack
Load Bearing Capacity (Metric) 200 kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.